
Nitric acid 60%, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Boiling Pt:∼120 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS05
Frequently Bought Together