Copper
Supplier: CHEMPUR
Jump to:
About this item
- CAS Number:7440-50-8
- Formula:Cu
- MDL Number:MFCD00010965
- Molecular Weight:63.55 g/mol
- Merck Index:13,02545
- Boiling Pt:2595 °C (1013 hPa)
- Melting Pt:1083 °C
- Density:8.94 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-159-6
4 Options Available Below
Product Details & Documents
- CAS Number:7440-50-8
- Formula:Cu
- MDL Number:MFCD00010965
- Molecular Weight:63.55 g/mol
- Merck Index:13,02545
- Boiling Pt:2595 °C (1013 hPa)
- Melting Pt:1083 °C
- Density:8.94 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-159-6
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



