Aluminium sulphate 0.3% in aqueous solution
Supplier: CHEMPUR
New Product
Certificates
About this item
CAS Number:10043-01-3
:
Formula:Al₂O₁₂S₃
MDL Number:MFCD00003423
:342.15 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:∼1 g/cm³ (20 °C)
EINECS:233-135-0
Frequently Bought Together