Potassium chloride 0.2 mol/l (0.2 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7447-40-7
:
Formula:KCl
MDL Number:MFCD00011360
:74.55 g/mol
Storage Temperature:Ambient
EINECS:231-211-8
Frequently Bought Together