Ammonia solution 0.1 mol/l (0.1 N) volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Storage Temperature:Ambient
EINECS:215-647-6
Frequently Bought Together