Nitric acid 6 mol/l (6 N) volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS05
Frequently Bought Together