Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nitric acid
Nitric acid 3%, pure analytical reagent
Nitric acid 3%, pure analytical reagent
CHMP115296051.1000
:
CHEMPUR
:
Nitric acid 3%, pure analytical reagent
CHMP115296051.1000
:
CHEMPUR
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
63.01 g/mol
:
Ambient
:
231-714-2
:
2031
:
8,II
:
7697-37-2
:
HNO₃
:
MFCD00011349