Order Entry
Poland
ContactUsLinkComponent
Nitric acid 1%, pure analytical reagent
Nitric acid 1%, pure analytical reagent
Catalog # CHMP115296042.1000
Supplier:  CHEMPUR
CAS Number:  
Nitric acid 1%, pure analytical reagent
Catalog # CHMP115296042.1000
Supplier:  CHEMPUR
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349