Triacetin (glycerol triacetate), pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:102-76-1
:
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716
:218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:4 °C
Density:1.1596 g/cm³ (20 °C)
Flash Pt:138 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-051-9
Frequently Bought Together