
Barium chloride 0.05 mol concentrated aqueous solution volumetric solution
Catalog # CHMP161579707.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
:208.24 g/mol
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-788-1
UN:3287
ADR:6.1,III
:GHS07
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445