

Cobalt(II) nitrate hexahydrate, pure
Supplier: CHEMPUR
Certificates
About this item
CAS Number:10026-22-9
:
Formula:Co(NO₃)₂·6H₂O
MDL Number:MFCD00149647
:291.03 g/mol
Merck Index:13,02469
Melting Pt:57 °C
Density:1.88 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-402-1
UN:1477
ADR:5.1,III
:GHS08
:GHS09
Frequently Bought Together