Manganese(II) carbonate, anhydrous, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:598-62-9
:
Formula:CMnO₃
MDL Number:MFCD00011116
:114.95 g/mol
Merck Index:13,05749
Density:3.12 g/cm³ (20 °C)
EINECS:209-942-9
Frequently Bought Together