Cobalt(II) chloride, anhydrous, pure analytical reagent
: CHEMPUR
:7646-79-9
:
:Cl₂Co
:MFCD00010938
:129.84 g/mol
:12,02498
:1049 °C (1013 hPa)
:735 °C
:1.6795 g/cm³ (20 °C)
:Refrigerator
:231-589-4
:3077
:9,III
:GHS08
:GHS07
:GHS09
Frequently Bought Together