
Barium bromide dihydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7791-28-8
:
Formula:BaBr₂·2H₂O
MDL Number:MFCD00149153
:333.17 g/mol
Merck Index:14,968
Melting Pt:850 °C
Density:3.58
Storage Temperature:Ambient
EINECS:234-140-0
UN:1564
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together