Trichloroacetic acid (TCA) 500 g/l in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
Synonyms: TCA
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
Boiling Pt:196
Melting Pt:54…56 °C
Density:≤1.07 g/cm³ (20 °C)
Molecular Weight:163.39 g/mol
Flash Pt:113
Storage Temperature:Ambient
UN:2564
ADR:8 II
Merck Index:13,09700
EINECS:200-927-2
:GHS05
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together