


Trichloroacetic acid (TCA) 30% in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:≤1.07 g/cm³ (20 °C)
EINECS:200-927-2
UN:2564
ADR:8,II
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together