


Mercury (II) oxide, red, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:21908-53-2
:
Formula:HgO
MDL Number:MFCD00011045
:216.59 g/mol
Merck Index:13,05908
Boiling Pt:356.6 °C (1013 hPa)
Melting Pt:> 400 °C
Density:11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:244-654-7
UN:1641
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Frequently Bought Together