
Sodium hydroxide 5% in aqueous solution, Technical Grade
Supplier: CHEMPUR
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Frequently Bought Together