Magnesium sulphate heptahydrate, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:10034-99-8
:
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
:246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2.66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
Frequently Bought Together