Chloroacetic acid, pure
: CHEMPUR
:79-11-8
:
:ClCH₂COOH
:MFCD00002683
:94.5 g/mol
:13,02129
:188 °C (1013 hPa)
:56…57 °C
:1.4043 g/cm³ (20 °C)
:126 °C
:Ambient
:201-178-4
:1751
:6.1,II
:GHS06
:GHS05
:GHS09
Frequently Bought Together