D-(-)-Fructose, pure
Supplier: CHEMPUR
Certificates
About this item
CAS Number:57-48-7
:
Formula:C₆H₁₂O₆
MDL Number:MFCD00148910
:180.16 g/mol
Merck Index:13,04295
Boiling Pt:552 °C (1013 hPa)
Melting Pt:100…110 °C
Density:1.59 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-333-3
Frequently Bought Together