
Ammonium iron(III) sulphate dodecahydrate, pure analytical reagent
Supplier: POCH AVANTOR BRAND
Certificates
About this item
CAS Number:7783-83-7
:
Formula:NH₄Fe(SO₄)₂·12H₂O
MDL Number:MFCD00150004
:482.2 g/mol
Merck Index:12,00549
Boiling Pt:230 °C (1013 hPa)
Melting Pt:39…41 °C
Density:1.71 g/cm³ (20 °C)
EINECS:233-382-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together