


Mercury (II) oxide, yellow, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:21908-53-2
:
Formula:HgO
MDL Number:MFCD00011045
:216.59 g/mol
Merck Index:13,05908
Melting Pt:500 °C
EINECS:244-654-7
UN:1641
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together