Mercury(I) chloride, pure analytical reagent
: CHEMPUR
:10112-91-1
:
:Cl₂Hg₂
:MFCD00011043
:472.09 g/mol
:12,05951
:384 °C (1013 hPa)
:383 °C
:7.15 g/cm³ (20 °C)
:Ambient
:233-307-5
:2025
:6.1,II
:GHS07
:GHS09
Frequently Bought Together