Erlenmeyer flask with screw cap, WHEATON®
Catalog # WHEA635134
Supplier: DWK Life Sciences
Specifications
- Thread:20-400
- Capacity:25 ml
- Environmentally Preferable:
- Pk:1
Specifications
About this item
These flasks, made of low extractable borosilicate glass, are used for continuous extraction of analytes from a solid into an organic solvent.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes