Order Entry
Poland
ContactUsLinkComponent
Decontamination Solution, DNA/RNA-ExitusPlus™
Decontamination Solution, DNA/RNA-ExitusPlus™
Supplier:  PanReac AppliChem
Certificates
undefined
Decontamination Solution, DNA/RNA-ExitusPlus™
Supplier:  PanReac AppliChem
Certificates

Specifications

  • Storage temperature:
    Ambient

Specifications

Activity test strips

Product Information