Order Entry
Poland
Orders LinkContactUsLinkComponent
Benzonase®

Benzonase®

Benzonase®
Certificates

2 Options Available Below

Product Details & Documents

Benzonase® is a highly active nuclease that degrades DNA and RNA to short oligonucleotides. It is used for the elimination of interfering DNA/RNA, for example at protein isolation, or before electrophoresis or chromatography.

Delivery information: Aluminium tube containing 500000 Units and Plastic bottle containing 100000 units.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1.01654.0001
  • 1.01656.0001
  • Enzyme name:
  • Benzonase®
  • Benzonase®

Specifications

Product Family Options

Product Information

  • Pk Info
    Pack type
    Pk
    Availability
    Price
  • 100,000 U/vial
    Plastic bottle
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT

    Specifications:
    More
    Specifications
    Cat. No.
    1.01654.0001
    Enzyme name
    Benzonase®
  • 500,000 U/vial
    Aluminium tube
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT

    Specifications:
    More
    Specifications
    Cat. No.
    1.01656.0001
    Enzyme name
    Benzonase®