Synthetic Poly-D-Lysine, Corning®
Catalog # 734-1102
Supplier: Corning
CAS Number:
Specifications
- Pk:20 mg
- Source:Synthetic
- Environmentally Preferable:Low Manufacturing Impact
- Storage conditions:Stable for at least three months at 2 to 8 °C (lyophilised) or one week at 2 to 8 °C (solubilised)
- Environmental certification:ISO 14001 - Bedford
- Protein synonyms:PDL
- Protein/peptide name:Poly-D-Lysine
- Purity:Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma
- Molecular weight:500-550 kD
- Formulation:Lyophilised from aqueous solution; reconstitute in distilled water
Specifications
About this item
Poly-D-lysine (PDL) is a synthetic molecule used as a coating to enhance cell attachment to plastic and glass surfaces. It has been used to culture a wide variety of cell types, particularly neurons, glial cells, and transfected cells.