Serum-free media
Catalog # FG0465
Supplier: Biochrom (part of Merck Millipore)
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 ml
- Description:Iscove's liquid medium with stable glutamine, with 3,024 g/l NaHCO₃, with 15 mg/l phenol red
- Seq:07