Order Entry
Poland
ContactUsLinkComponent
TISAB III concentrate (Total Ionic Strength Adjustment Buffer) for fluoride determinations
TISAB III concentrate (Total Ionic Strength Adjustment Buffer) for fluoride determinations
Catalog # REGETISAF5
Supplier:  REAGECON
CAS Number:  
TISAB III concentrate (Total Ionic Strength Adjustment Buffer) for fluoride determinations
Catalog # REGETISAF5
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    TISAB III concentrate for fluoride determinations
  • pH:
    5,0 - 5,5

Specifications

About this item