VWR® Traceable®, Digital Dial Thermometers
Supplier: VWR Collection
Certificates
About this item
These thermometers are safer than mercury thermometers. The smooth probe (620-2024) is suitable for viscous solutions or semi-solids, thanks to its piercing tip.
- Readings updated every second
- Large, bright LCD for easy reading
- 620-2082 with metal disc (Ø: 12,7 mm) for surface measurements
Stainless steel probe (Lר: 133×3,8 mm).
Delivery: Supplied with silver-oxide battery and Traceable certificate of calibration.