9-Fluorenylmethyl chloroformate (Fmoc chloride), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:28920-43-6
:
Formula:C₁₅H₁₁ClO₂
MDL Number:MFCD00001138
:258.7 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:249-313-6
UN:2923
Frequently Bought Together