Order Entry
Poland
ContactUsLinkComponent
9-Fluorenylmethyl chloroformate (Fmoc chloride) 98%
9-Fluorenylmethyl chloroformate (Fmoc chloride) 98%
Certificates
undefined
9-Fluorenylmethyl chloroformate (Fmoc chloride) 98%
Certificates

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    60 to 64 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together