(-)-Shikimic acid for synthesis, Sigma-Aldrich®
: Merck
:138-59-0
:
:C₇H₁₀O₅
:MFCD00066278
:174.15 g/mol
:M-Clarity™ quality level = MQ200
:205-334-2
:GHS05
:Ambient
- Solubility:180 g/l (20 °C)
- Melting Point:190 °C
- Molar Mass:174.15 g/mol
- Synonyms:(3R,4S,5R)-3,4,5-Trihydroxy-1-cyclohexenecarboxylic acid
- Hill Formula:C7H10O5
Frequently Bought Together