Order Entry
Poland
ContactUsLinkComponent
 
Pyruvaldehyde (methylglyoxal) 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL67028-100ML
Supplier:  Merck
CAS Number:  
Pyruvaldehyde (methylglyoxal) 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL67028-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
 :72.06 g/mol
EINECS:201-164-8
CAS Number:78-98-8
Formula:C₃H₄O₂
MDL Number:MFCD00006960