Order Entry
Poland
ContactUsLinkComponent
2-(Chloromethyl)-1,3-dioxolane, Sigma-Aldrich®
2-(Chloromethyl)-1,3-dioxolane, Sigma-Aldrich®
Catalog # SIAL329991-25G
Supplier:  Merck
CAS Number:  
2-(Chloromethyl)-1,3-dioxolane, Sigma-Aldrich®
Catalog # SIAL329991-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :122.55 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:219-908-5
CAS Number:2568-30-1
Formula:C₄H₇ClO₂
MDL Number:MFCD00043402