Order Entry
Poland
ContactUsLinkComponent
tert-Butyllithium 1.6-3.2 mol/l in heptane, Sigma-Aldrich®
tert-Butyllithium 1.6-3.2 mol/l in heptane, Sigma-Aldrich®
Catalog # SIAL94439-100ML-F
Supplier:  Merck
CAS Number:  
tert-Butyllithium 1.6-3.2 mol/l in heptane, Sigma-Aldrich®
Catalog # SIAL94439-100ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :64.06 g/mol
EINECS:209-831-5
CAS Number:594-19-4
Formula:C₄H₉Li
MDL Number:MFCD00008795