3-(Methylthio)butanal 95%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 95% CAS No.: 16630-52-7 Molecular Formula: CH3SCH(CH3)CH2CHO Formula Weight: 118.20 Flash Point: 62°(144°F) Boiling Point: 63-65°/11mm MDL No.: MFCD00036622
Synonyms: 3-(Methylthio)butyraldehyde
- CAS Number:16630-52-7
- Formula:C₅H₁₀OS
- MDL Number:MFCD00036622
- Molecular Weight:118.2 g/mol
- Boiling Pt:63…65 °C (11 mmHg)
- Flash Pt:62 °C (144 °F)
- Storage Temperature:Ambient
- EINECS:240-678-7
2 Options Available Below
Product Details & Documents
Purity: 95% CAS No.: 16630-52-7 Molecular Formula: CH3SCH(CH3)CH2CHO Formula Weight: 118.20 Flash Point: 62°(144°F) Boiling Point: 63-65°/11mm MDL No.: MFCD00036622
Synonyms: 3-(Methylthio)butyraldehyde
- CAS Number:16630-52-7
- Formula:C₅H₁₀OS
- MDL Number:MFCD00036622
- Molecular Weight:118.2 g/mol
- Boiling Pt:63…65 °C (11 mmHg)
- Flash Pt:62 °C (144 °F)
- Storage Temperature:Ambient
- EINECS:240-678-7
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



