Order Entry
Poland
ContactUsLinkComponent
D-(-)-Ornithine hydrochloride, Sigma-Aldrich®
D-(-)-Ornithine hydrochloride, Sigma-Aldrich®
Catalog # SIALO5250-1G
Supplier:  Merck
CAS Number:  
D-(-)-Ornithine hydrochloride, Sigma-Aldrich®
Catalog # SIALO5250-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.62 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:240-729-3
CAS Number:16682-12-5
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00012917