DL-Ornithine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1069-31-4
:
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00065398
:168.62 g/mol
Melting Pt:291 °C (dec.)
Storage Temperature:Ambient
EINECS:213-956-0
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1.5 %
- Solubility:(10 % in water) Clear colorless to light yellow solution
- Titration with HClO4:98.5 to 101.5 %
Specification Test Results
Frequently Bought Together