
Citric acid monohydrate 30% in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:5949-29-1
:
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
:210.14 g/mol
EINECS:201-069-1
:GHS05
Storage Temperature:Ambient
Frequently Bought Together