4,6-Dichloro-5-nitropyrimidine
Catalog # MOLEM69992794
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:193.98 g/mol
EINECS:224-340-6
CAS Number:4316-93-2
Formula:C₄HCl₂N₃O₂
MDL Number:MFCD00006107