


Copper(II) nitrate hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13778-31-9
:
Formula:CuN₂O₆·H₂O
MDL Number:MFCD00149669
:205.57 g/mol
ADR:5.1,II
:M-Clarity™ quality level = MQ100
EINECS:221-838-5
UN:1477
:GHS03
:GHS07
:GHS09
Frequently Bought Together