Zinc chloride 5% in aqueous solution, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
:136.3 g/mol
Storage Temperature:Ambient
EINECS:231-592-0
Frequently Bought Together