
Calcium chloride, anhydrous, pure analytical reagent
Supplier: POCH AVANTOR BRAND
Certificates
About this item
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Merck Index:14,01659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2.174 g/cm³ (20 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
Frequently Bought Together