Order Entry
Poland
Orders LinkContactUsLinkComponent
Benzaldehyde, pure analytical reagent

Benzaldehyde, pure analytical reagent

Irritant
Supplier:   CHEMPUR
Benzaldehyde, pure analytical reagent
Supplier:  CHEMPUR
Certificates
Benzaldehyde, pure analytical reagent
Benzaldehyde, pure analytical reagent

4 Options Available Below

Product Details & Documents
  • CAS Number:100-52-7
  • Formula:C₆H₅CHO
  • MDL Number:MFCD00003299
  • Molecular Weight:106.12 g/mol
  • Merck Index:13,01057
  • Boiling Pt:179 °C (1013 hPa)
  • Melting Pt:–26 °C
  • Density:1.049 g/cm³ (20 °C)
  • Flash Pt:64 °C
  • Storage Temperature:Ambient
  • EINECS:202-860-4
  • UN:1990
  • ADR:9,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 65,50 zł
    65,50 zł
    1 * 100 mL
       65,50 zł

  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 136 zł
    136 zł
    1 * 250 mL
       136 zł

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 198 zł
    198 zł
    1 * 500 mL
       198 zł

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 333 zł
    333 zł
    1 * 1 L
       333 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.