Order Entry
Poland
Orders LinkContactUsLinkComponent
Citric acid monohydrate 40% in aqueous solution

Citric acid monohydrate 40% in aqueous solution

Corrosive
Supplier:   CHEMPUR
Citric acid monohydrate 40% in aqueous solution
Supplier:  CHEMPUR
Certificates
Citric acid monohydrate 40% in aqueous solution
Citric acid monohydrate 40% in aqueous solution

3 Options Available Below

Product Details & Documents
Synonyms:  2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
  • CAS Number:5949-29-1
  • Formula:C₆H₈O₇.H₂O
  • MDL Number:MFCD00149972
  • Molecular Weight:210.14 g/mol
  • Storage Temperature:Ambient
  • EINECS:201-069-1
Product Family Options

Product Information