

4-Methyl-2-pentanone, specially pure
CHMP114879509.1000
: CHEMPUR
:
- Pack type:Glass bottle
- Pk:1 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
:
:100.16 g/mol
:13,05227
:117.4 °C (1013 hPa)
:–84 °C
:0.801 g/cm³ (20 °C)
:16 °C
:Ambient
:203-550-1
:1245
:3,II
:GHS02
:GHS07
:108-10-1
:(CH₃)₂CHCH₂COCH₃
:MFCD00008938