

4-Methyl-2-pentanone, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:108-10-1
:
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938
:100.16 g/mol
Merck Index:13,05227
Boiling Pt:117.4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.801 g/cm³ (20 °C)
Flash Pt:16 °C
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Frequently Bought Together