Butyl acetate, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:123-86-4
:
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445
:116.16 g/mol
Merck Index:13,01534
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–76 °C
Density:0.875 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:204-658-1
UN:1123
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Frequently Bought Together